3-Hydrazinobenzoesäure ≥99%

Lieferant: ABCR

Synonyme: 3-Hydrazinylbenzoic acid

AB131443.0050
ABCRAB131443.0050EA 0 EUR
ABCRAB131443.0050
3-Hydrazinobenzoesäure ≥99%
3-Hydrazinobenzoic acid

Purity: 99% CAS No.: 38235-71-1 Molecular Formula: C7H8N2O2 Formula Weight: 152.15 Melting Point: ca 194° dec. MDL No.: MFCD00191447

Formel: C₇H₈N₂O₂
Molecular Weight: 152,15 g/mol
Schmelzpunkt: ∼194 °C (Zersetzung)
MDL: MFCD00191447
CAS-Nummer: 38235-71-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR