Methyl-3,4-dimethoxybenzoat ≥98%

Lieferant: Thermo Scientific

Synonyme: 3,4-Dimethoxybenzoesäuremethylester, Methylveratrumat

B23568.18 AB131937.0050
B23568.18EA 50.9 EUR
B23568.18 ABCRAB131937.0050
Methyl-3,4-dimethoxybenzoat ≥98%
Methyl-3,4-dimethoxybenzoat

Purity: 98+% CAS No.: 2150-38-1 Molecular Formula: C10H12O4 Formula Weight: 196.20 Melting Point: 57-61° Boiling Point: 283° MDL No.: MFCD00008430

Formel: C₁₀H₁₂O₄
Molecular Weight: 196,2 g/mol
Siedepunkt: 283 °C
Schmelzpunkt: 57…61 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00008430
CAS-Nummer: 2150-38-1
EINECS: 218-424-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR