Bromacetylchlorid

Lieferant: ABCR

AB138071.0500
ABCRAB138071.0500EA 0 EUR
ABCRAB138071.0500
Bromacetylchlorid
Bromacetylchlorid
Formel: BrCH₂COOCl
Molecular Weight: 157,39 g/mol
MDL: MFCD00000724
CAS-Nummer: 22118-09-8
EINECS: 244-790-7
UN: 3265
ADR: 8,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR