1,1,2-Trichlorpropan

Lieferant: ABCR

Synonyme: 2-Chloropropylidene chloride

AB251723.0005
ABCRAB251723.0005EA 0 EUR
ABCRAB251723.0005
1,1,2-Trichlorpropan
1,1,2-Trichloropropane
Formel: Cl₂HCCHClCH₃
Molecular Weight: 147,43 g/mol
MDL: MFCD00018864
CAS-Nummer: 598-77-6
EINECS: 209-951-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR