Methyl-3,4,5-trihydroxybenzoat ≥98%

Lieferant: Thermo Scientific
Achtung

Synonyme: 3,4,5-Trihydroxybenzoesäuremethylester, Methylgallat, Gallussäuremethylester

L05926.18 L05926.30 L05926.0250
L05926.18EA 49.3 EUR
L05926.18 L05926.30 ABCRL05926.0250
Methyl-3,4,5-trihydroxybenzoat ≥98%
Methyl-3,4,5-trihydroxybenzoat

Purity: 98% CAS No.: 99-24-1 Molecular Formula: C8H8O5 Formula Weight: 184.15 Melting Point: 198-203° Beilstein Reference: 2113180 Merck Reference: 14,4345 MDL No.: MFCD00002194

Formel: C₈H₈O₅
Molecular Weight: 184,15 g/mol
Siedepunkt: 450 °C (1013 hPa)
Schmelzpunkt: 198…203 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00002194
CAS-Nummer: 99-24-1
EINECS: 202-741-7
Merck Index: 14,4345

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR