Antifade solution
Lieferant: Abnova
U0032
ABNOU0032EA 196 EUR
ABNOU0032
Antifade solution
Antifade solution
Antifade Solution is comprised of Antifade and glycerol in an aqueous buffer solution.
  • for Fluorescent In Situ Hybridization (Cell)