Ammoniumeisen(II)sulfat Hexahydrat 98%, reinst
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
Ferrous Ammonium sulphate Hexahydrate, di-Ammonium iron (II) sulphate hexahydrate, Ferrous ammonium sulphate hexahydrate, Ammonium iron(Ⅱ) sulphate hexahydrate, Ammonium ferrous sulphate hexahydrate, Ammonium iron(II) sulphate hydrate
193250050
ACRO193250050EA
128
EUR
ACRO193250050
Ammoniumeisen(II)sulfat Hexahydrat 98%, reinst
Ammonium iron (II) sulphate hexahydrate
Formel:
(NH₄)₂Fe(SO₄)₂·6H₂O Molecular Weight: 392,14 g/mol Schmelzpunkt: 100 °C (Zers.) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00150530 CAS-Nummer: 7783-85-9 |
Spezifikation Testergebnisse
Appearance | Green to blue-green to blue to dark blue Powder |
pH | 3 to 5 |
Titration with KMnO4 | ≥97.5 % |
Insoluble matter | ≤0.05 % (in dilute H2SO4) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...