Chrysoidine, rein
Lieferant: Thermo Scientific
|
Gefahr
|
200740500
ACRO200740500EA
47.3
EUR
ACRO200740500
Chrysoidine, rein
Chrysoidine
Appearance: Bordeaux to purple Crystalline powder
Formel:
C₁₂H₁₃ClN₄ Molecular Weight: 248,714 g/mol Schmelzpunkt: 235 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00012976 CAS-Nummer: 532-82-1 UN: 3077 ADR: 9,III |
Spezifikation Testergebnisse
Appearance | Bordeaux to purple Crystalline powder |
Infrared spectrum | Conforms |
UV spectrum | Complies |
Visual transition interval | from pH 4.0 (orange) to pH 7.0 (yellow) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...