Mitomycin C (2 mg/48 mg NaCl)
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
Mitomycin, Ametycin
226940020
ACRO226940020EA
133
EUR
ACRO226940020
Mitomycin C (2 mg/48 mg NaCl)
Mitomycin C
Formel:
C₁₅H₁₈N₄O₅ Molecular Weight: 334,33 g/mol Schmelzpunkt: >360 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00078109 CAS-Nummer: 50-07-7 UN: 2811 ADR: 6.1,III |
Spezifikation Testergebnisse
Appearance | Blue to violet Powder |
HPLC | ≥99 % with regard to the non-formulated material |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...