Cresyl violet perchlorate 98%, rein

Lieferant: Thermo Scientific
Achtung

Synonyme: Cresyl violet perchlorate, laser grade, 5-iminobenzo[a]phenoxazin-9-amine, perchloric acid

405775000
ACRO405775000EA 211 EUR
ACRO405775000
Cresyl violet perchlorate 98%, rein
Cresyl violet perchlorate

Appearance: Green Powder

Formel: C₁₆H₁₂ClN₃O₅
Molecular Weight: 361,738 g/mol
Schmelzpunkt: 331…333 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00012671
CAS-Nummer: 41830-80-2

Bestellen

Spezifikation Testergebnisse

Appearance Green Powder
Infrared spectrum Conforms
HPLC ≥97.5 %
UV Lambda max 590 to 596 nm (in Methanol)
Molar absorption coefficient 67400 l/(mol.cm) (at Lambda max)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR