Methyl orange-xylene cyanol-Lösung in Wasser, rein

Lieferant: Thermo Scientific

414675000
ACRO414675000EA 61.9 EUR
ACRO414675000
Methyl orange-xylene cyanol-Lösung in Wasser, rein
Methyl orange-xylene cyanol

Appearance: Dark blue-green Solution

Formel: C₃₉H₄₁N₅Na₂O₁₀S₃
Dichte: 1 g/cm³
Lagertemperatur: Raumtemperatur
MDL: MFCD00077269
CAS-Nummer: 62758-15-0

Bestellen

Spezifikation Testergebnisse

Appearance Dark blue-green Solution
Visual transition interval from pH 3.2 (purple) to pH 4.2 (green)
GC ≥88 % may occur as tautomers

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR