Dental clamp forcep, Aesculap®
Lieferant: AESCULAP AG
DP152R
AESCDP152REA 0 EUR
AESCDP152R
Dental clamp forcep, Aesculap®
Pinzetten
This angled combination clamp forcep is particularly suitable for positioning brackets in the posterior teeth.
  • Mouth parts fluted
  • Spindle on forcep end is used for adjusting the brackets