NM522 competent cells
Lieferant: AGILENT
200233
AGLS200233EA 114 EUR
AGLS200233
NM522 competent cells
Kompetente Zellen
NM522 competent cells are recombination-proficient and EcoK restriction-deficient.
  • Recombination-proficient and EcoK restriction-deficient
  • Blue-white colour screening possible for recombinant plasmids
  • Transformation efficiency >1×10⁸ transformants/µg DNA
NM6522 competent cells are a classic E. coli strain for economical cloning when you don’t need supercompetent efficiencies. NM522 is a recombination-proficient and EcoK restriction deficient host strain. Intrinsic antibiotic resistance aids cell selection after a complete and successful transformation.
Bitte beachten For research use only. Not for use in diagnostic procedures.