IPTG (Isopropyl-β-D-thiogalactopyranosid) ≥99% (durch TLC)
Lieferant: AGILENT
|
Achtung
|
Synonyme:
IPTG
300127
AGLS300127EA
126
EUR
AGLS300127
IPTG (Isopropyl-β-D-thiogalactopyranosid) ≥99% (durch TLC)
IPTG (Isopropyl-β-D-thiogalactopyranosid)
IPTG is for use in combination with X-gal to detect β-galactosidase activity in blue/white selection assays.
Bitte beachten For Research Use Only. Not for Use in Diagnostic Procedures.
Formel:
C₉H₁₈O₅S Molecular Weight: 238,31 g/mol Schmelzpunkt: 112,..114 °C Lagertemperatur: Gefrierschrank |
MDL:
MFCD00063273 CAS-Nummer: 367-93-1 |
Spezifikation Testergebnisse
Purity | ≥99% (TLC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...