Protein Broad Range P240 Kit
Lieferant: AGILENT
5191-6640
AGLS5191-6640EA 1900 EUR
AGLS5191-6640
Protein Broad Range P240 Kit
Analysatoren Protein-Analysatoren
Automate the CE-SDS analysis of denatured proteins with capillary electrophoresis. Protein broad range P240 kit, 275 samples. ProteoAnalyzer system kit for the analysis of diverse proteins through 240 kDa.
  • Quick analysis - separate 12 samples in parallel in approximately 30 minutes
  • Short preparation time - prepare samples with only 10 minutes hands-on time
  • Broad quantitative range - allow 2 to 2000 ng/µl input sample concentration
  • Variable sample types - consistently separate difficult proteins such as membrane proteins and crude lysates
  • High resolution - clearly resolve glycosylated and non-glycosylated isoforms
  • No system peak interference - covalent labeling method eliminates comigrating system peaks from the separation
  • Multiple applications - analyse samples under both reducing and non-reducing conditions
The Agilent Protein Broad Range P240 kit is developed to provide a solution for diverse proteins including monoclonal antibodies, soluble fractions, membrane fractions, and crude lysates through 240 kDa. Complete rejuvenation of the capillary electrophoresis environment between runs ensures high consistency for protein analysis.
The Protein Broad Range P240 kit provides high resolution results for protein separations from 10 to 240 kDa in 30 minutes. This allows for analysis of varied sample sizes in a single streamlined automated electrophoresis run. The rapid covalent labeling method avoids system peaks appearing in the analysis region while providing a 3-log dynamic range allowing for impurity detection down to 0,1%.
Lieferumfang: Includes reagents necessary for running 275 samples.
Bitte beachten For research use only. Not for use in diagnostic procedures.