Cooking thermometer
Lieferant: ALLA ATELIERS
801.060-GB
ALLA801.060-GBEA 36.3 EUR
ALLA801.060-GB
Cooking thermometer
Thermometer Analoge Thermometer
Internal scale glass thermometer: −10 to +60 °C , for baking, milk and wine.
  • White polypropylene alimentary protection sleeve