Hair hygrometer
Lieferant: AMARELL, ARNO
LF105562
AMARLF105562EA 81.98 EUR
AMARLF105562
Hair hygrometer
Hygrometer Hygrometer, analog
This hair hygrometer is used for precise measuring of relative humidity.
  • Round shape