Sleeve protectors, vinyl
Lieferant: ANSELL HEALTH CARE
VSB-8-HS
ANSEVSB-8-HSEA 46.2 EUR
ANSEVSB-8-HS
Sleeve protectors, vinyl
Einweg-Kleidung
These protective sleeves are made of vinyl. They provide excellent protection against chemicals, snags, punctures and abrasions.
  • Unsupported
  • Elasticated at both ends
Thickness: 0,203 mm.