HRP - conjugate stabiliser, SeramunStab® STC
Lieferant: Apollo Scientific
BIL3115-100ML
APOSBIL3115-100MLEA 65.4 EUR
APOSBIL3115-100ML
HRP - conjugate stabiliser, SeramunStab® STC
Proteinkonjugate und Markierungen
Liquid product red odourless pH-value 7,2 to 7,4 boiling point 103 °C. Optimised for background reduction.