Ponceau S

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-Hydroxy-4-(2-sulpho-4-[4-sulphophenylazo]phenylazo)-2,7-naphthalenedisulphonic acid sodium salt, CI 27195, Ponceau, 3-Hydroxy-4-((2-sulpho-4-((4-sulphophenyl)diazenyl)phenyl)diazenyl)naphthalene-2,7-disulphonic acid, tetrasodium salt

BIP1405-25G
APOSBIP1405-25GEA 98.6 EUR
APOSBIP1405-25G
Ponceau S
Ponceau S
Formel: C₂₂H₁₂N₄Na₄O₁₃S₄
Molecular Weight: 760,56 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00003892
CAS-Nummer: 6226-79-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR