3-Hydroxypicolinsäure

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-Hydroxypyridine-2-carboxylic acid

OR14330-1G
APOSOR14330-1GEA 21.9 EUR
APOSOR14330-1G
3-Hydroxypicolinsäure
3-Hydroxypicolinsäure
Formel: C₆H₅NO₃
Molecular Weight: 139,11 g/mol
Schmelzpunkt: 213…219 °C (Zers.)
Lagertemperatur: Raumtemperatur
MDL: MFCD00006294
CAS-Nummer: 874-24-8
EINECS: 212-859-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR