7-Bromisochinolin

Lieferant: Apollo Scientific
Achtung

OR40468-250MG
APOSOR40468-250MGEA 21.9 EUR
APOSOR40468-250MG
7-Bromisochinolin
7-Bromoisoquinoline
Formel: C₉H₆BrN
Molecular Weight: 208,0600 g/mol
Schmelzpunkt: 67…72 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD07368661
CAS-Nummer: 58794-09-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR