3-Brom-2-methylpropansäure 95%
Lieferant: Apollo Scientific
Synonyme:
Captopril EP Impurity D, 3-Bromo-2-methylpropanoic acid, 3-Bromo-2-methylpropionic Acid
OR921957-25G
APOSOR921957-25GEA
109
EUR
APOSOR921957-25G
3-Brom-2-methylpropansäure 95%
3-Brom-2-methylpropansäure
Formel:
C₄H₇BrO₂ Molecular Weight: 167,00 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD01310530 CAS-Nummer: 56970-78-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...