4-Bromoisoquinoline-6-carboxylic acid ≥98%

Lieferant: Apollo Scientific

OR935026-100MG
APOSOR935026-100MGEA 102 EUR
APOSOR935026-100MG
4-Bromoisoquinoline-6-carboxylic acid ≥98%
4-Bromoisoquinoline-6-carboxylic acid
Formel: C₁₀H₆BrNO₂
Molecular Weight: 252,064 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28502758
CAS-Nummer: 1637280-23-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR