Ammoniumeisen(II)sulfat 0.1 M (0.1 N) volumetrische Lösung
Lieferant: PanReac AppliChem
Synonyme:
Ferroammoniumsulfat, Eisen(II)ammoniumsulfat
181369.1611
APPC181369.1611EA
62
EUR
APPC181369.1611
Ammoniumeisen(II)sulfat 0.1 M (0.1 N) volumetrische Lösung
Ammoniumeisen(II)sulfat
Iron(II) ammonium sulphate can be used for the determination of COD, using ferroin indicator.
Formel:
H₈FeN₂O₈S₂ Molecular Weight: 284,05 g/mol |
MDL:
MFCD00051039 CAS-Nummer: 10045-89-3 UN: 2796 ADR: 8,II |
Spezifikation Testergebnisse
Presentation | Clear, Light green Liquid |
Solubility | Miscible with water (D 20/4) 1,045 |
Titer at 20 °C | 0,999 - 1,001 |
Traceability | NIST |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...