Human RHOT1 ELISA Kit
Lieferant: Aviva Systems Biology
OKEH03665
AVIVOKEH03665EA 1040 EUR
AVIVOKEH03665
Human RHOT1 ELISA Kit
Tests ELISAs
Aviva Systems Biology RHOT1 ELISA Kit (Human) (OKEH03665) is based on standard sandwich enzyme-linked immuno-sorbent assay technology. An antibody specific for RHOT1 has been pre-coated onto a 96-well plate (12×8 Well Strips) and blocked. Standards or test samples are added to the wells and removed. A biotinylated detector antibody specific for RHOT1 is added, incubated and followed by washing. Avidin-Peroxidase Conjugate is then added, incubated and unbound conjugate is washed away. An enzymatic reaction is produced through the addition of TMB substrate which is catalyzed by HRP generating a blue color product that changes yellow after adding acidic stop solution. The density of yellow coloration read by absorbance at 450 nm is quantitatively proportional to the amount of sample RHOT1 captured in well.
  • Mean recovery when spiking into sample matrices at concentrations within the dynamic range: 97%.
Mean Intra-assay CV%: <= 7.4% (n = 20)
Mean Inter-assay CV%: <= 9.2% (n = 20)