Calciumchlorid-Lösung 0.5 M für die Molekularbiologie

Lieferant: Aviva Systems Biology

Synonyme: Chlorcalcium

OORA00231
AVIVOORA00231EA 185 EUR
AVIVOORA00231
Calciumchlorid-Lösung 0.5 M für die Molekularbiologie
Calciumchlorid

0.5 M Calcium Chloride is suitable in molecular biology assays such as genetic transfection.


This product was aseptically filtered through a Millipore 0.22 micron filter into clean, pre-sterilized containers. The product was tested on trypticase soy agar for 24 hours, 48 hours and 72 hours and was found to be negative for bacteria.


This product is a 0.5M concentrated stock solution and should be diluted appropriately with distilled, deionized water or equivalent to its final working concentration. Store container at room temperature (18 to 26°C). Expiration date is six (6) months from date of opening container.

Formel: CaCl₂
Molecular Weight: 110,98 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00010903
CAS-Nummer: 10043-52-4
EINECS: 233-140-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR