Bromphenol Blue–Xylene Cyanole-Lösung, Sigma-Aldrich®
Lieferant: SIGMA ALDRICH MICROSCOPY
|
Gefahr
|
B3269-5ML
B3269-5MLEA
63
EUR
B3269-5ML
Bromphenol Blue–Xylene Cyanole-Lösung, Sigma-Aldrich®
Bromphenol Blue–Xylene Cyanole
Bromophenol Blue–Xylene Cyanol Dye solution has been used in the formamide loading buffer for the size purification of ligated RNA from polyacrylamide gel.