Coomassie Brillantblau G 250 (Konzentrat), Sigma-Aldrich®
Lieferant: SIGMA ALDRICH MICROSCOPY
|
Gefahr
|
Synonyme:
Hydrogen[4-[4-(p-ethoxyanilino)-4'-[ethyl(m-sulfonatobenzyl)amino]-2'-methylbenzhydrylen]-3-methylcyclohexa-2,5-dien-1-yliden](ethyl)(m-sulfonatobenzyl)ammonium Mononatriumsalz, Coomassie Blau G 250, Brillantindocyanin G, Acid Blue 90, C.I. 42655, Brillantblau G
Brilliant Blue G is a methanol based stain which is especially designed for staining protein in polyacrylamide gels, but will also stain protein in agarose gels (IEF). The stain contains methanol and acetic acid so gels do not require fixing prior to staining. Water is added directly to the bottle to produce 1 liter of ready-to-use staining solution.
Formel:
C₄₇H₄₈N₃NaO₇S₂ Molecular Weight: 854,04 g/mol Flammpunkt: 9,7 °C (geschlossener Tiegel) |
MDL:
MFCD00078482 CAS-Nummer: 6104-58-1 UN: 3286 ADR: 3 (6.1, 8),II |
Spezifikation Testergebnisse
Suitability for staining protein in polyacrylamide and agarose gels | Pass |
Appearance (Colour) | Dark Blue |
Appearance (Form) | Liquid |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...