Coomassie® Brillantblau R 250 in Methanol für SDS-PAGE, Sigma-Aldrich®

Lieferant: SIGMA ALDRICH MICROSCOPY
Gefahr

Synonyme: Brillantblau R, Hydrogen[4-[4-(p-ethoxyanilino)-4'-[ethyl(m-sulfonatobenzyl)amino]benzhydrylen]cyclohexa-2,5-dien-1-yliden](ethyl)(m-sulfonatobenzyl)ammonium Mononatriumsalz, Coomassie Blau R 250, Xylenbrillantcyanin, Serva Blue R, Brillantindocyanin 6B, Acid Blue 83

M-Clarity™ Qualitätsstufe = MQ200

B8647-1EA
B8647-1EAEA 118 EUR
B8647-1EA
Coomassie® Brillantblau R 250 in Methanol für SDS-PAGE, Sigma-Aldrich®
Coomassie® Brillantblau R 250

Brilliant Blue R concentrate is a methanol based stain especially designed for staining protein in polyacrylamide but will also stain agarose gels. The stain contains methanol and acetic acid so gels do not require fixing prior to staining. Brilliant Blue R Concentrate has been used for protein band detection after nondenaturing gradient gel electrophoresis, isoelectric focussing and SDS-PAGE (sodium dodecyl sulfate polyacrylamide gel electrophoresis). For detection of protein bands following electrophoresis.

Formel: C₄₅H₄₄N₃NaO₇S₂
Molecular Weight: 825,98 g/mol
Flammpunkt: 14 °C (geschlossener Tiegel)
Lagertemperatur: Room Temperature
MDL: MFCD00041762
CAS-Nummer: 6104-59-2
UN: 1230
ADR: 3 (6.1),II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c31af4d351c996cbdc924f4cd417f5a79130e517&vid=ff2c7d54c1b28238b558e1b01338fc40dcf6c595&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Appearance (Colour) Dark Blue
Appearance (Form) Liquid
Suitability, Suitable for use as a protein stain in electrophoresis Pass

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR