Probe for digital thermometer, CoolProbe™
Lieferant: BIOCISION
BCS-200
BCISBCS-200EA 252 EUR
BCISBCS-200
Probe for digital thermometer, CoolProbe™
Temperaturfühler
Traceable temperature monitors for accurate temperature monitoring of samples while in CoolRack or CoolSink modules. Universal probe for use with other sample tubes and plates.
  • LCD digital display with minimum, maximum and current temperature display
  • Min/max memory, high/low alarm
  • Sampling rate of fast (10 seconds) or normal (60 seconds)
Standards: NIST traceable