BIT 9500 serum substitute
Lieferant: BIOCONCEPT
5-18S02-H
BCON5-18S02-HEA 217 EUR
BCON5-18S02-H
BIT 9500 serum substitute
Zellkulturmedien
Replaces FBS/FCS/serum in all cell culture media including various media for stem cells, primary cells and cell lines.
  • Eliminates Lot-to-Lot variability inherent in FBS/FCS/all forms of serum
  • Eliminates the need to test and reserve FBS/FCS/serum
  • Eliminates the need for inactivation
  • Eliminates the viral /bacterial/fungal contamination risk associated with FBS/FCS/serum
  • Reduces the price of media enrichment
  • Supports growth of all cells including stem cells, primary cells, 3D cultures etc
Serum is a highly variable non-standardized product requiring extensive testing to find the batch compatible with your cell types. The composition of serum-free medium on the other hand is well defined and ensures lot-to-lot consistency. BIT 9500 serum substitute replaces serum for the culture of cells and was developed for use in serum-free culture conditions with defined composition. BIT 9500 contains bovine serum albumin (BSA), insulin and transferrin (BIT) in Iscove’s IMDM. For BSA we use tested batches of EDQM certified Probumin™ BSA
Species: Human and mouse
Standards: EDQM certified; ISO 9001; cGMP.