Jar, PP, with screw cap, SP Bel-Art
Lieferant: Bel-Art Products, a Part of SP
H179100000
BELAH179100000EA 82.5 EUR
BELAH179100000
Jar, PP, with screw cap, SP Bel-Art
Dosen Dosen, Universal
White PP jar with screw cap in an assortment of sizes and is used for storage of liquid or dry stock, or for use with samples.
Screw cap with PE foam lining and are not autoclavable.