SP Bel-Art sidENTRY™, Glove box
Lieferant: Bel-Art Products, a Part of SP
H500260000
BELAH500260000EA 8740 EUR
BELAH500260000
SP Bel-Art sidENTRY™, Glove box
Handschuhboxen und -beutel
This small and portable glove box is designed for remote and field use. It is manufactured from transparent acrylic plastic (6,35 mm thick).
  • Glove box stability is assured by four adjustable levelers that also level the box and all its contents
  • Stopcocks are included ready for assembly
  • Includes large gasketed door on side of unit
  • Features two 20,32 cm glove ports compatible with standard glove box gloves