Culture tubes, with screw cap
Lieferant: BELLCO GLASS
2012-16150
BELC2012-16150EA 547 EUR
BELC2012-16150
Culture tubes, with screw cap
Röhrchen Kulturröhrchen
Manufactured from type 1 class B borosilicate glass, with 15-415 black phenolic rubber lined screw cap.
  • Very good chemical resistance
  • Very good temperature resistance