Centrifuge tubes with seal caps
Lieferant: BIOTIX
2570.X 2571.X 2576.X
BITX2570.XEA 0 EUR
BITX2570.X BITX2571.X BITX2576.X
Centrifuge tubes with seal caps
Röhrchen Zentrifugenröhrchen
PP, clear.
  • Excellent chemical resistance
  • Excellent temperature resistance
  • Printed
With flat top rim seal cap.