Microtubes with screw cap
Lieferant: BIOTIX
3733.S.X 3744.S.X
BITX3733.S.XEA 455 EUR
BITX3733.S.X BITX3744.S.X
Microtubes with screw cap
Röhrchen Mikroröhrchen
PP, with PE cap
  • Leak proof seal to –80 °C
  • DNase, RNase and pyrogen free
  • Centrigugation 20000×g for 15 minutes
  • Autoclavable
Ideal for centrifugation to PCR and freezer storage.