Rat Recombinant Hip (from E. coli)
Lieferant: BIOMOL RESEARCH LABORATORIES
SPP-767E
BMOLSPP-767EEA
0
EUR
BMOLSPP-767E
Rat Recombinant Hip (from E. coli)
Proteine und Peptide
Hsc70-interacting protein (Hip) is a cytosolic protein that exists as a homo-oligomer and participates in the regulation of the heat shock protein Hsc70 in eukaryotic cells.
- Produced in E. coli
- Purified by multi-step chromatography (SDS-PAGE, western blot)
It has a liquid formulation in 20 mM MOPS at pH 7,2, containing 50 mM sodium chloride and 2 mM magnesium chloride.