Double bosshead, cross type
Lieferant: BOCHEM
5333
BOCH5333EA 13.5 EUR
BOCH5333
Double bosshead, cross type
Klemmen Stativmuffen
Cross type bossheads made of aluminium.
  • For rods up to 16,5 mm
  • Angle: 90°
  • With thumb screw (M8)