Thermometer for distillation processes, BOLA
Lieferant: Bohlender
B290-03
BOHLB290-03EA 198 EUR
BOHLB290-03
Thermometer for distillation processes, BOLA
Thermometer Analoge Thermometer
Distillation thermometer made of PTFE jacketed glass. Suitable for 500 to 6000 ml reaction vessels.