Fluoroplastic (PTFE) spray
Lieferant: Bohlender
H958-04
BOHLH958-04EA 28 EUR
BOHLH958-04
Fluoroplastic (PTFE) spray
Schmiermittel
Release and anti-blocking agent as well as lubricant for laboratory. Excellent gliding and separating effects.
  • Dry and greaseless
  • CFC- and silicone-free
  • Heat resistant up to +260°C
  • Non-adhesive and dirt-repellent
  • Physiologically safe