Desiccator cabinet, black, SICCO Star
Lieferant: Bohlender
V1891-07
BOHLV1891-07EA 1200 EUR
BOHLV1891-07
Desiccator cabinet, black, SICCO Star
Exsikkatoren Trockenkammern
Cabinet has a reinforced aluminium frame, lightproof panels made of black acrylic glass which prevent light incidence and protect against ultraviolet radiation. Specially designed for the storage of substances to the exclusion of light transmission and ultraviolet radiation.
  • Door with magnetic catch and circular rubber seal
  • Easy to read electronic hygrometer
  • Stackable telescopic shelves
  • Temperature resistance from −20 to +70 °C
This cabinet is designed for storing or drying humidity sensitive products using silica gel. The tight fitting door protects the contents from contamination from the atmosphere. The controlled environment inside the desiccators is ideal for storing reference materials, retained samples, metallographic specimen, tobacco and DNA-samples.
Lieferumfang: Supplied with four shelves made of acrylic glass, tray, hygrometer and desiccant, usable with up to 26 shelves.