Glove box, SICCO
Lieferant: Bohlender
V1984-08
BOHLV1984-08EA 4180 EUR
BOHLV1984-08
Glove box, SICCO
Handschuhboxen und -beutel
This PMMA glove box allows safe working with sensitive compounds in a controlled environment.
  • Robust and light-weight aluminium construction
  • Superb all-round visibility
  • Non slip rubber feet
  • Cable lead-ins for electric devices
  • Large front opening
  • Connectors for gas-filling and aeration including appropriate tubing
Aluminium frame with panels made of acrylic glass, base panel made of PE. Front panel with two glove ports, two lead-ins for cables on the back side (upper left and upper right). Work space including two hose connectors for gas-filling and aeration and appropriate tubing (1×5 m tubing for gas-filling made of PVC, 1×5 m aeration tubing made of PE).
Hinged lid opening: 850×425 mm
Lieferumfang: Supplied with one pair of natural rubber gloves (size 9).