Printer cleaning kit
Lieferant: Brady
018556
BRDY018556EA 16.07 EUR
BRDY018556
Printer cleaning kit
Wischtücher Reinigungskits und -Pads
This cleaning kit for printers includes: Instruction sheet, five cleaning cards soaked in IPA and the material safety data sheet.