Lockout Bags
Lieferant: Brady
065780
BRDY065780EA 41.41 EUR
BRDY065780
Lockout Bags
Beutel Aufbewahrungsbeutel
Lockout bag made of rugged canvas with drawstrings.
  • Can be locked closed
  • Legend: LOCK OUT FOR SAFETY
For securing plugs on portable applications, cables on pneumatic connectors, crane pendants.