Brady® Alignment Roll for BradyJet™ J7300 Colour Label Printer
Lieferant: Brady
176579
BRDY176579EA 180 EUR
BRDY176579
Brady® Alignment Roll for BradyJet™ J7300 Colour Label Printer
Farbbänder
Use after replacing the ink cartridge for printhead alignment sequences.
  • For use with the BradyJet™ J7300 colour label printer