Brady® M410 Magnetic Accessory
Lieferant: Brady
176991
BRDY176991EA 78.13 EUR
BRDY176991
Brady® M410 Magnetic Accessory
Drucker
The printer magnet attachment gives the convenient option to attach your label printer to panels, I-beams and any other magnetic-receptive surface during use or storage.
  • Designed for use with the M410 label printer from Brady®
  • Allows printer to affix to panel door or metal surface for more hands-free operation or storage
  • Attaches to back of printer with screws to ensure stability
The printer magnet attachment gives the convenient option to attach your label printer to panels, I-beams and any other magnetic-receptive surface during use or storage. Designed for use with the M410 label printer from Brady®. Allows printer to affix to panel door or metal surface for more hands-free operation or storage. Attaches to back of printer with screws to ensure stability.
Bestellinformation: Order reference M410-MAGNET.
Lieferumfang: Package includes the printer magnet attachment only (printer sold separately).