Signs, floor signs, wash your hands, Brady®
Lieferant: Brady
306901
BRDY306901EA 19.23 EUR
BRDY306901
Signs, floor signs, wash your hands, Brady®
Warn- und Verbotszeichen
Signs are made of surface printed anti-skid laminated white polyester with a permanent acrylic pressure-sensitive rubber adhesive. They are overlaminated with a clear anti-slip vinyl film.
  • Anti-slip safety sign
  • Circular shape, white on blue
  • Ideal for floormarking applications in stairs, hallways, and offices
Thickness: 0,254 mm.
Standards: Tested according DIN 51130:2014 with a slip resistance classification of R9.