ISO Safety Signs - Warning; Substance or Mixture that can Cause an Environmental Hazard
Lieferant: Brady
309879
BRDY309879EA 6.06 EUR
BRDY309879
ISO Safety Signs - Warning; Substance or Mixture that can Cause an Environmental Hazard
Warn- und Verbotszeichen
ISO safety sign depicts 'Warning; Substance or Mixture that can Cause an Environmental Hazard' message. It helps to avoid any possibility to have contact with or spills of substances or mixtures considered hazardous to health.
  • Triangle shape
  • Black on yellow finish
  • 0,099 mm thickness
  • Permanent acrylic adhesive and glossy finish
Applications includes facility identification, hazard communication identification.
Standards: Compliant with Directive 2011/65/EU and 2015/863.