Nordic ID Stix UHF RFID Reader + Writer (USB) EU
Lieferant: Brady
311095
BRDY311095EA 485.45 EUR
BRDY311095
Nordic ID Stix UHF RFID Reader + Writer (USB) EU
Barcode- und RFID-Systeme
Extra small UHF RFID reader + writer with USB connectivity.
  • Suitable for Android devices
  • Easy installation
  • USB powered
  • No battery
This Stix is an affordable credit card sized UHF RFID reader and writer with USB connectivity. It allows an easy way of bringing RFID capabilities into your PC or laptop.